Order Entry
ContactUsLinkComponent
Mouse Recombinant ICAM2
Mouse Recombinant ICAM2
# de catalogue 75791-712
Fournisseur:  Prosci
Numéro CAS:  
Mouse Recombinant ICAM2
# de catalogue 75791-712
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    CD102 antigen|ICAM-2|CD102|Icam2|Intercellular adhesion molecule 2|Lymphocyte function-associated AG-1 counter-receptor
  • Protein/peptide name:
    ICAM2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    49.7 kD
  • Sequence:
    Lys23-Gln222
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM Tris,150mM NaCl,pH8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Cat. no.:
    75791-712
  • Supplier No.:
    92-575

Spécifications

A propos de cet article

ICAM-2 is a 55-65 kD transmembrane glycoprotein possessing 2 extracellular Ig domains, a single transmembrane domain, and a short 26-amino acid cytoplasmic domain. ICAM-2 is expressed on most leukocytes, and is strongly expressed on vascular endothelial cells. Interactions of ICAM-2 and the integrin receptors mediate cell adhesion in a wide range of lymphocyte, monocyte, natural killer cell, and granulocytewith other cells, and play important roles in many adhesion-dependent immune and inflammation responses, such as T cell aggregation, NK-cell cytotoxicity and migration, lymphocyte recirculation, etc. Serum levels of ICAM-2 correlated significantly with the inflammatory and course sequences of trichinosis in mice and had a similar relation with blood eosinophilia. So, estimation of ICAM-2 serum levels may prove useful in diagnosis of trichinosis recent infections, and in monitoring the prognosis and response to treatment.

  • Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought