Order Entry
ContactUsLinkComponent
Mouse Recombinant BMP Receptor IA
Mouse Recombinant BMP Receptor IA
# de catalogue 75791-380
Fournisseur:  Prosci
Numéro CAS:  
Mouse Recombinant BMP Receptor IA
# de catalogue 75791-380
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Tag sequence:
    C-Fc-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    ALK-3|ALK-3/Bone morphogenetic protein receptor type-1A/BMP type-1A receptor/BMPR-1A/Activin receptor-like kinase 3/BMP-2/BMP-4 receptor/Serine/threonine-protein kinase receptor R5/SKR5/CD292/Bmpr1a/Bmpr/Acvrlk3|CD292
  • Protein/peptide name:
    BMP Receptor IA
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    42.2 kD
  • Sequence:
    Gln24-Arg152
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75791-380
  • Supplier No.:
    92-405

Spécifications

A propos de cet article

ALK-3 is a type I receptor for bone morphogenetic proteins (BMPs) which belong to the protein kinase superfamily, TKL Ser/Thr protein kinase family and TGFB receptor subfamily. The BMP receptors consists of the type I receptors BMPR1A and BMPR1B and the type I I receptor BMPR2. Seven known type I serine/threonine kinases and five mammalian type II serine/threonine kinase receptors function in TGF-beta superfamily signal transduction. The downstream molecules of the type I BMP receptors include the Smad (Smad1, 5 and 8) proteins that are phosphorylated in a ligand-dependent manner, and relay the BMP signal from the receptors to target genes in the nucleus. Type II receptors phosphorylate and activate type I receptors which autophosphorylate, then bind and activate SMAD transcriptional regulators. ALK-3 contains a GS domain and a protein kinase domain. ALK-3 is widely expressed. Defects in BMPR1A gene are a cause of a significant proportion of cases of Juvenile polyposis syndrome (JPS).

  • Fusion-Tag: C-Fc-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought