Order Entry
ContactUsLinkComponent
Human Recombinant UBE2D1 (from E. coli)
Human Recombinant UBE2D1 (from E. coli)
# de catalogue 75791-138
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant UBE2D1 (from E. coli)
# de catalogue 75791-138
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    N-GST tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    Ubiquitin-conjugating enzyme E2(17)KB 1|Ubiquitin-protein ligase D1|UBCH5A|UBC4/5 homolog|Ubiquitin carrier protein D1|UbcH5|SFT|UBC5A|Ubiquitin-conjugating enzyme E2 D1|Stimulator of Fe transport|Ubiquitin-conjugating enzyme E2-17 kDa 1
  • Protein/peptide name:
    UBE2D1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    42.9 kD
  • Sequence:
    Met1-Met147
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 50mM HEPES,150mM NaCl,2mM DTT,10% Glycerin pH7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75791-138
  • Supplier No.:
    92-272

Spécifications

A propos de cet article

Ubiquitin-conjugating enzyme E2 D1(UBE2D1)belongs to the ubiquitin-conjugating enzyme family. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This enzyme is closely related to a stimulator of iron transport (SFT), and is up-regulated in hereditary hemochromatosis. It also functions in the ubiquitination of the tumor-suppressor protein p53 and the hypoxia-inducible transcription factor HIF1alpha by interacting with the E1 ubiquitin-activating enzyme and the E3 ubiquitin-protein ligases.

  • Fusion-Tag: N-GST tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought