Order Entry
ContactUsLinkComponent
Human Recombinant UFM1 activating enzyme (from E. coli)
Human Recombinant UFM1 activating enzyme (from E. coli)
# de catalogue 75791-050
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant UFM1 activating enzyme (from E. coli)
# de catalogue 75791-050
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    UBE1DC1|Ubiquitin-like modifier-activating enzyme 5|Ubiquitin-activating enzyme E1 domain-containing protein 1|UFM1-activating enzyme|UBA5|ThiFP1
  • Protein/peptide name:
    UFM1 activating enzyme
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    47 kD
  • Sequence:
    Met1-Met404
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl pH 8.0,1mM DTT, 10% glycerol, 50mM NaCl. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75791-050
  • Supplier No.:
    92-228

Spécifications

A propos de cet article

UBA5 is a member of the ubiquitin-activating E1 family and UBA5 subfamily. Ubiquitin and ubiquitin-like proteins are recognized as covalently conjugated to various cellular substrates by a three-step enzymatic pathway. The ubiquitin-activating enzyme (E1) has a vital role in the first step of ubiquitination pathway to activate ubiquitin or ubiquitin-like proteins. UBA5 activates ubiquitin-fold modifier 1, a ubiquitin-like post-translational modifier protein, via the formation of a high-energy thioester bond. UBA5 is located primarily in cytoplasm, while it generally localizes to the nucleus in presence of SUMO2.

  • Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought