Order Entry
ContactUsLinkComponent
Human Recombinant B Lymphocyte Kinase (from E. coli)
Human Recombinant B Lymphocyte Kinase (from E. coli)
# de catalogue 75790-808
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant B Lymphocyte Kinase (from E. coli)
# de catalogue 75790-808
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    B Lymphocyte Kinase
  • Enzyme synonyms:
    Tyrosine-Protein Kinase Blk, B Lymphocyte Kinase, p55-Blk, BLK
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    58.7 kD
  • Sequence:
    Gly2-Pro505
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM Tris, 500 mM NaCl, 1 mM DTT, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-808
  • Supplier No.:
    92-107

Spécifications

A propos de cet article

Tyrosine-Protein Kinase Blk (BLK) contains one protein kinase domain, one SH2 domain and one SH3 domain

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

BLK is a non-receptor tyrosine kinase, which is involved in B-lymphocyte development, differentiation and signaling. B-cell receptor (BCR) signaling requires a tight regulation of several protein tyrosine kinases and phosphatases, and associated coreceptors. Signaling through BLK plays an important role in transmitting signals through surface immunoglobulines and supports the pro-B to pre-B transition, as well as the signaling for growth arrest and apoptosis downstream of B-cell receptor. Defects in BLK are a cause of maturity-onset diabetes of the young type 11 (MODY11).

Customers Who Bought This Also Bought