Order Entry
ContactUsLinkComponent
Human Recombinant CCL11 (from E. coli)
Human Recombinant CCL11 (from E. coli)
# de catalogue 75790-794
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant CCL11 (from E. coli)
# de catalogue 75790-794
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    C-C Motif Chemokine 11|Eosinophil Chemotactic Protein|SCYA11|Small-Inducible Cytokine A11|CCL11|Eotaxin
  • Protein/peptide name:
    CCL11
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    8.56 kD
  • Sequence:
    Gly24-Pro97
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-794
  • Supplier No.:
    92-100

Spécifications

A propos de cet article

C-C Motif Chemokine 11 (CCL11) is a secreted protein that belongs to the intercrine beta (chemokine CC) family. In response to the presence of allergens, CCL11 selectively recruits eosinophils, a prominent feature of allergic inflammatory reactions. The effects of CCL11 are mediated by its binding to a G-protein-linked receptor known as a chemokine receptor. Chemokine receptors for CCL11 include CCR2, CCR3 and CCR5. However, it has been found that CCL11 has high degree selectivity for its receptor, such that they are inactive on neutrophils and monocytes, which do not express CCR3.

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought