Order Entry
ContactUsLinkComponent
Human Recombinant UBE2I (from E. coli)
Human Recombinant UBE2I (from E. coli)
# de catalogue 75790-766
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant UBE2I (from E. coli)
# de catalogue 75790-766
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    N-GST tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    UBCE9|UBE2I|Ubiquitin-Conjugating Enzyme E2 I|Ubiquitin Carrier Protein I|Ubiquitin Carrier Protein 9|SUMO-Protein Ligase|SUMO-Conjugating Enzyme UBC9|Ubiquitin-Protein Ligase I|UBC9|p18
  • Protein/peptide name:
    UBE2I
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    44.4 kD
  • Sequence:
    Met1-Ser158
  • Formulation:
    Supplied as a 0.2 um filtered solution of 50mM HEPES, 150mM NaCl, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    used for biological assays
  • Cat. no.:
    75790-766
  • Supplier No.:
    92-086

Spécifications

A propos de cet article

SUMO-Conjugating Enzyme UBC9 (UBC9) belongs to the ubiquitin-conjugating enzyme family. UBC9 is homologous to ubiquitin-conjugating enzymes (E2s). However, instead of conjugating ubiquitin, UBC9 conjugates a ubiquitin homologue, Small Ubiquitin-Like Modifier 1 (SUMO-1). The conjugation of ubiquitin requires the activities of ubiquitin-activating (E1) and conjugating (E2) enzymes. It is suggested that UBC9 might play a role in DNA repair and perhaps even in aging.

  • Fusion-Tag: N-GST tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought