Order Entry
ContactUsLinkComponent
Human Recombinant DFF45 (from E. coli)
Human Recombinant DFF45 (from E. coli)
# de catalogue 75790-696
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant DFF45 (from E. coli)
# de catalogue 75790-696
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    H13|DFF-45|Inhibitor of CAD|DFFA|ICAD|DNA Fragmentation Factor 45 kDa Subunit|DFF1|DNA Fragmentation Factor Subunit Alpha|DFF45
  • Protein/peptide name:
    DFF45
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    38.7 kD
  • Sequence:
    Met1-Thr331
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    used for biological assays
  • Cat. no.:
    75790-696
  • Supplier No.:
    92-051

Spécifications

A propos de cet article

DNA Fragmentation Factor Subunit Alpha (DFFA). DFFA exists as a heterodimer (DFF) with DFFB. DFF is activated once DFFA is cleaved by Caspase-3. The cleaved fragments of DFFA detach from DFFB (the active component of DFF), which in turn triggers DNA fragmentation as well as chromatin condensation during apoptosis. A reduced level of DFFA detected in ovarian endometriosis may be a part of an apoptosis-resistant mechanism enhancing the disease progression.

  • Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought