Order Entry
ContactUsLinkComponent
Human Recombinant BMP Receptor IA (from Cells)
Human Recombinant BMP Receptor IA (from Cells)
# de catalogue 75790-496
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant BMP Receptor IA (from Cells)
# de catalogue 75790-496
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-Fc-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    BMP Type-1A Receptor|BMPR-1A|BMPR1A|SKR5|ALK-3|Activin Receptor-like Kinase 3|ALK3|Bone Morphogenetic Protein Receptor Type-1A|CD292|ACVRLK3|Serine/Threonine-Protein Kinase Receptor R5
  • Protein/peptide name:
    BMP Receptor IA
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    42.1 kD
  • Sequence:
    Gln24-Arg152
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-496
  • Supplier No.:
    91-930

Spécifications

A propos de cet article

Bone Morphogenetic Protein Receptor Type-1A (BMPR1A) belongs to the TKL Ser/Thr protein kinase family and TGFB receptor subfamily, including the type I receptors BMPR1A and BMPR1B and the type II receptor BMPR2. BMPR1A is a single-pass type I membrane protein and highly expressed in skeletal muscle. BMPR1A contains one GS domain and one protein protein kinase domain. BMPR1A is necessary for the extracellular matrix depostition by osteoblasts. BMPR1A can activate SMAD transcriptional regulators, binding with ligands. Defects in BMPR1A are a cause of juvenile polyposis syndrome, Cowden disease and hereditary mixed polyposis syndrome 2 (HMPS2).

  • Fusion-Tag: C-Fc-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought