Order Entry
ContactUsLinkComponent
Human Recombinant Endothelial cell adhesion molecule (from Cells)
Human Recombinant Endothelial cell adhesion molecule (from Cells)
# de catalogue 75790-440
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant Endothelial cell adhesion molecule (from Cells)
# de catalogue 75790-440
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    ESAM|Endothelial Cell-Selective Adhesion Molecule
  • Protein/peptide name:
    Endothelial cell adhesion molecule
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    50.8 kD
  • Sequence:
    Gln30-Ala247
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-440
  • Supplier No.:
    91-902

Spécifications

A propos de cet article

Endothelial Cell Adhesion Molecule (ESAM) is a 55 kDa type I transmembrane glycoprotein member of the JAM family of immunoglobulin superfamily molecules. The 390 amino acid Human ESAM contains a 216 amino acid extracellular domain (ECD) with a V-type and a C2-type immunoglobulin (Ig) domain. ESAM is specifically expressed at endothelial tight junctions and on activated platelets and performs homophilic adhesion activity. The adaptor protein membrane-associated guanylate kinase MAGI-1 has been identified as an intracellular binding partner of ESAM. In addition, ESAM at endothelial tight junctions participates in the migration of neutrophils through the vessel wall, possibly by influencing endothelial cell contacts. ESAM-deficient mice were described with lowered angiogenic potential, and accordingly, overexpression of ESAM is closely associated with certain tumor growth and metastasis. ESAM is expressed on endothelial cells, activated platelets and megakaryocytes. The ECD of human and mouse ESAM share 69% amino acid identity.

  • Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought