To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme name:Galactokinase 2
- Enzyme synonyms:N-acetylgalactosamine kinase, also known as GalNAc kinase, Galactokinase 2, GALK2 and GK2, is a member of the GHMP kinase family and GalK subfamily., GALK2, GK2
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:51.4 kD
- Sequence:Met1-Ala458
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75790-240
- Supplier No.:91-794
Spécifications
A propos de cet article
GALK2 acts as a galactokinase when galactose is present at high concentrations
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
GALK2 may be involved in a salvage pathway for the reutilization of free GalNAc derived from the degradation of complex carbohydrates. GALK2 has been reported to participate in pathways , such as Amino sugar and nucleotide sugar metabolism, Galactose metabolism and Metabolic pathways.