Order Entry
ContactUsLinkComponent
Mouse Recombinant Proprotein Convertase 9, ProSci
Mouse Recombinant Proprotein Convertase 9, ProSci
# de catalogue 75790-218
Fournisseur:  Prosci
Numéro CAS:  
Mouse Recombinant Proprotein Convertase 9, ProSci
# de catalogue 75790-218
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Enzyme type:
    Recombinant
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    Proprotein Convertase 9
  • Enzyme synonyms:
    Proprotein Convertase Subtilisin/Kexin Type 9, Neural Apoptosis-Regulated Convertase 1, NARC-1, Proprotein Convertase 9, PC9, Subtilisin/Kexin-Like Protease PC9, Pcsk9, Narc1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    74.8 kD
  • Sequence:
    Gln35-Gln692
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 50 mM HEPES, 150 mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-218
  • Supplier No.:
    91-783

Spécifications

A propos de cet article

Proprotein Convertase Subtilisin/Kexin Type 9 (PCSK9) is a secretory subtilase belonging to the proteinase K subfamily

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

PCSK9 is synthesized as a soluble zymogen that undergoes autocatalytic intramolecular processing in the ER , the pro domain and mature chain secrete together through noncovalent interactions. PCSK9 binds with low-density lipoprotein receptor (LDLR) and plays a major regulatory role in cholesterol homeostasis.PCSK9 also plays a role in neural development.

Customers Who Bought This Also Bought