To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme name:Hyaluronidase-1
- Enzyme synonyms:Hyaluronidase-1, Hyal-1, Hyaluronoglucosaminidase-1, Lung Carcinoma Protein 1, LuCa-1, HYAL1, LUCA1
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:47.18 kD
- Sequence:Phe22-Trp435
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, 10% Glycerol, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75790-208
- Supplier No.:91-778
Spécifications
A propos de cet article
Hyaluronidase-1 (HYAL1) is a secreted lysosomal hyaluronidase that belongs to the glycosyl hydrolase 56 family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
HYAL1 contains one EGF-like domain and is highly expressed in the liver, kidney, and heart, but it is weakly expressed in the lung, placenta, and skeletal muscle. HYAL1 is thought to be involved in cell proliferation, migration, and differentiation. It may play a role in promoting tumor progression and blocking the TGFB1-enhanced cell growth. Mutations in HYAL1 are associated with mucopolysaccharidosis type IX, or hyaluronidase deficiency.