Order Entry
ContactUsLinkComponent
Human Recombinant Prostaglandin-D2 Synthase (from Cells)
Human Recombinant Prostaglandin-D2 Synthase (from Cells)
# de catalogue 75790-166
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant Prostaglandin-D2 Synthase (from Cells)
# de catalogue 75790-166
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    Prostaglandin-D2 Synthase
  • Enzyme synonyms:
    Prostaglandin-H2 D-Isomerase, Beta-Trace Protein, Cerebrin-28, Glutathione-Independent PGD Synthase, Lipocalin-Type Prostaglandin-D Synthase, Prostaglandin-D2 Synthase, PGD2 Synthase, PGDS, PGDS2, PTGDS, PDS
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    19.7 kD
  • Sequence:
    Ala23-Gln190
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, 10% Glycerol, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-166
  • Supplier No.:
    91-757

Spécifications

A propos de cet article

Prostaglandin-H2 D-Isomerase (PTGDS) belongs to the Lipocalin family of calycin superfamily

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

PTGDS is preferentially expressed in the brain. PTGDS catalyzes the conversion of PGH2 to PGD2, a prostaglandin involved in smooth muscle contraction/relaxation and a potent inhibitor of platelet aggregation. PTGDS is involved in a variety of CNS functions, such as sedation, REM sleep and PGE2-induced allodynia, and may have an anti-apoptotic role in oligodendrocytes. PTGDS binds small non-substrate lipophilic molecules and may act as a scavenger for harmful hydrophopic molecules and a secretory retinoid and thyroid hormone transporter. It possibly participates in development and maintenance of the blood-brain, blood-retina, blood-aqueous humor, blood-testis barrier, the central nervous system and male reproductive system.

Customers Who Bought This Also Bought