Order Entry
ContactUsLinkComponent
Human Recombinant Tripeptidyl-Peptidase I (from Cells)
Human Recombinant Tripeptidyl-Peptidase I (from Cells)
# de catalogue 75790-142
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant Tripeptidyl-Peptidase I (from Cells)
# de catalogue 75790-142
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    Tripeptidyl-Peptidase I
  • Enzyme synonyms:
    Tripeptidyl-Peptidase 1, TPP-1, Cell Growth-Inhibiting Gene 1 Protein, Lysosomal Pepstatin-Insensitive Protease, LPIC, Tripeptidyl Aminopeptidase, Tripeptidyl-Peptidase I, TPP-I, TPP1, CLN2
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    60.35 kD
  • Sequence:
    Ser20-Pro563
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl,1 mM GaCl₂, 10% Glycerol, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-142
  • Supplier No.:
    91-745

Spécifications

A propos de cet article

Tripeptidyl-Peptidase 1 (TPP1) belongs to the peptidase S53 family

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

TPP1 is detected in all tissues examined with highest levels in heart and placenta and relatively similar levels in other tissues. TPP1 is lysosomal serine protease with tripeptidyl-peptidase I activity. TPP1 may act as a non-specific lysosomal peptidase which generates tripeptides from the breakdown products produced by lysosomal proteinases. TPP1 requires substrates with an unsubstituted N-terminus. TPP1 mutations have also been shown to cause neuronal ceroid lipofuscinosis type 2 (CLN2).

Customers Who Bought This Also Bought