ContactUsLinkComponent
Human Recombinant Ribonuclease 3 (from Cells)
Human Recombinant Ribonuclease 3 (from Cells)
# de catalogue 75790-096
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant Ribonuclease 3 (from Cells)
# de catalogue 75790-096
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    Ribonuclease 3
  • Enzyme synonyms:
    Eosinophil Cationic Protein, ECP, Ribonuclease 3, RNase 3, RNASE3, RNS3
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    16.6 kD
  • Sequence:
    Arg28-Ile160
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl,1 mMDTT, 10% Glycerol,pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-096
  • Supplier No.:
    91-718
Spécifications
A propos de cet article
Ribonuclease 3 (RNASE3) is a basic protein that is localized to the eosinophil primary matrix and belongs to the pancreatic ribonuclease family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
RNASE3 is released during degranulation of eosinophils. RNASE3 possesses a wide variety of biological activities. RNASE3 interacts with bacterial lipopolysaccharide (LPS) and lipoteichoic acid (LTA). RNASE3 exhibits antibacterial activity, including cytoplasmic membrane depolarization of preferentially Gram-negative, but also Gram-positive strains. It promotes E. coli outer membrane detachment, alteration of the overall cell shape and partial loss of cell content.
Customers Who Bought This Also Bought