Order Entry
ContactUsLinkComponent
Human Recombinant Pyruvate Kinase (from Cells)
Human Recombinant Pyruvate Kinase (from Cells)
# de catalogue 75790-062
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant Pyruvate Kinase (from Cells)
# de catalogue 75790-062
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    Pyruvate Kinase
  • Enzyme synonyms:
    Pyruvate Kinase Isozymes R/L, Pyruvate Kinase 1, R-Type/L-Type Pyruvate Kinase, Red Cell/Liver Pyruvate Kinase, PKLR, PK1, PKL
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    62.9 kD
  • Sequence:
    Met1-Ser574
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM Tris-HCl, 1 mM DTT 0.2 M NaCl, 10% glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-062
  • Supplier No.:
    91-701

Spécifications

A propos de cet article

Pyruvate Kinase Isozymes R/L (PKLR) belongs to the pyruvate kinase family, There are 4 isozymes of pyruvate kinase in mammals: L, R, M1 and M2

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

L type is major isozyme in the liver; R is found in red cells; M1 is the main form in muscle, heart and brain; M2 is found in early fetal tissues. PKLR exists as a homotetramer and catalyzes the production of phosphoenolpyruvate from pyruvate and ATP. Defects in PKLR are also the cause of pyruvate kinase deficiency of red cells, which is a frequent cause of hereditary non-spherocytic hemolytic anemia.

Customers Who Bought This Also Bought