To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme name:Sorbitol Dehydrogenase
- Enzyme synonyms:Sorbitol Dehydrogenase, L-Iditol 2-Dehydrogenase, SORD
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:39.3 kD
- Sequence:Ala2-Pro357
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl,0.2 M NaCl,5mM DTT, 20% glycerol,pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75790-002
- Supplier No.:91-671
Spécifications
A propos de cet article
Sorbitol dehydrogenase, also known as L-iditol 2-dehydrogenase and SORD, is a member of the zinc-containing alcohol dehydrogenase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
SORD exsits in a homotetramer and binds one zinc ion per subunit. SORD is expressed in kidney and epithelial cells of both benign and malignant prostate tissue. SORD can converts sorbitol to fructose and catalyzes the interconversion of polyols and their corresponding ketoses, and together with aldose reductase to make up the sorbitol pathway. SORD is up-regulated by androgens and down-regulated by castration. SORD may play a role in the sperm motility by providing an energetic source for sperm.