Order Entry
ContactUsLinkComponent
Human Recombinant UBE2B (from Cells)
Human Recombinant UBE2B (from Cells)
# de catalogue 75789-990
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant UBE2B (from Cells)
# de catalogue 75789-990
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    HR6B|Ubiquitin-Conjugating Enzyme E2-17 kDa|UBE2B|RAD6B|Ubiquitin Carrier Protein B|Ubiquitin-Protein Ligase B|hHR6B|Ubiquitin-Conjugating Enzyme E2 B|RAD6 Homolog B
  • Protein/peptide name:
    UBE2B
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    18.3 kD
  • Sequence:
    Met1-Ser152
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-990
  • Supplier No.:
    91-665

Spécifications

A propos de cet article

Ubiquitin-Conjugating Enzyme E2 B (UBE2B) is a member of the ubiquitin-conjugating enzyme family. UBE2B accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. It is shown that UBE2B interacts with RAD18, UBR2, and WAC. UBE2B is required for post-replicative DNA damage repair. Additional, UBE2B plays a role in sepsis-induced muscle protein proteolysis and cancer-induced cachexia.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought