Order Entry
ContactUsLinkComponent
Human Recombinant Transaldolase (from Cells)
Human Recombinant Transaldolase (from Cells)
# de catalogue 75789-972
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant Transaldolase (from Cells)
# de catalogue 75789-972
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    Transaldolase
  • Enzyme synonyms:
    Transaldolase, TALDO1, TAL, TALDO, TALDOR
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    38.5 kD
  • Sequence:
    Met1-Lys337
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 10% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-972
  • Supplier No.:
    91-656

Spécifications

A propos de cet article

Transaldolase (TALDO1) belongs to the transaldolase family of Type 1 subfamily

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

TALDO1 is expressed selectively in oligodendrocytes of the brain. TALDO1 is a key enzyme of the nonoxidative pentose phosphate pathway providing ribose-5-phosphate for nucleic acid synthesis and NADPH for lipid biosynthesis. This pathway can also maintain glutathione at a reduced state and thus protect sulfhydryl groups and cellular integrity from oxygen radicals. TALDO1 deficiency results in telangiectases of the skin, hepatosplenomegaly and enlarged clitoris.

Customers Who Bought This Also Bought