To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Storage conditions:Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme name:Pancreatic Lipase-Related 1
- Enzyme synonyms:Pancreatic Lipase-Related Protein 1, PL-RP1, PNLIPRP1, PLRP1
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:50.99 kD
- Sequence:Lys18-Cys467
- Formulation:Lyophilized from a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75789-812
- Supplier No.:91-568
Spécifications
A propos de cet article
Pancreatic Lipase-Related Protein 1 (PNLIPRP1) belongs to the Lipase family within the AB hydrolase superfamily
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
PNLIPRP1 is a secreted protein and contains one PLAT domain. PNLIPRP1 is involved in lipid metabolic process, acting as a negative regulator of pancreatic lipase activity by competing with pancreatic lipase for colipase occupancy. PNLIPRP1 may play a role in inhibiting dietary triglyceride digestion, but it lacks detectable lipase activity towards triglycerides, diglycerides, phosphatidylcholine, galactolipids or cholesterol esters.