Order Entry
ContactUsLinkComponent
Human Recombinant Proteinase 3 (from Cells)
Human Recombinant Proteinase 3 (from Cells)
# de catalogue 75789-760
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant Proteinase 3 (from Cells)
# de catalogue 75789-760
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    Proteinase 3
  • Enzyme synonyms:
    Myeloblastin, AGP7, C-ANCA Antigen, Leukocyte Proteinase 3, PR-3, PR3, Neutrophil Proteinase 4, NP-4, P29, Wegener Autoantigen, PRTN3, MBN
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    25.64 kD
  • Sequence:
    Ala26-Arg249
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 10 mM TrisHCl, 150 mM NaCl, 10% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-760
  • Supplier No.:
    91-542

Spécifications

A propos de cet article

Proteinase-3 is a neutral serine proteinase that belongs to the peptidase S1 family and Elastase subfamily

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

It contains one peptidase S1 domain and it is expressed mainly in neutrophil granulocytes. The primary function of Proteinase-3 is thought to be degradation of extracellular proteins at sites of inflammation, but excessive or prolonged proteolytic activity may cause harmful effects in the body. It is the epitope of anti-neutrophil cytoplasmic antibodies (ANCAs) of the cANCA (cytoplasmic subtype) class, a type of antibody frequently found in the disease Wegener's granulomatosis.

Customers Who Bought This Also Bought