Order Entry
ContactUsLinkComponent
Human Recombinant VSIG4 (from Cells)
Human Recombinant VSIG4 (from Cells)
# de catalogue 75789-674
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant VSIG4 (from Cells)
# de catalogue 75789-674
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    Z39IG|CRIg|Protein Z39Ig|V-Set and Immunoglobulin Domain-Containing Protein 4|VSIG4
  • Protein/peptide name:
    VSIG4
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    30.2 kD
  • Sequence:
    Arg20-Pro283
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological Assays
  • Cat. no.:
    75789-674
  • Supplier No.:
    91-499

Spécifications

A propos de cet article

V-Set and Immunoglobulin Domain-Containing Protein 4 (VSIG4) is a 45-50 kDa macrophage-specific transmembrane glycoprotein that belongs to the B7 family-related protein and an Ig superfamily member. In contrast to the B7 family members which contain two IgG domains, VSIG4 contains one complete V-type Ig domain and a truncated C-type I g domain. VSIG4 is abundantly expressed in several fetal tissues. In adult tissues, the highest expression of VSIG4 is in lung and placenta. It is also expressed in resting macrophages. No VSIG4 expression appears to be present in T and B cells. The specific expression of VSIG4 on resting macrophages in tissue suggests that this inhibitory ligand may be important for the maintenance of T cell unresponsiveness in healthy tissues. VSIG4 functions as a negative regulator of T cell activation, and may be involved in the maintenance of peripheral T cell tolerance, and is also identified as a potent suppressor of established inflammation. VSIG4 is a phagocytic receptor, strong negative regulator of T-cell proliferation and IL2 production. It is a potent inhibitor of the alternative complement pathway convertases. Human VSIG4 is 399 amino acids (aa) in length. It is a type I transmembrane (TM) glycoprotein that contains a 264 aa extracellular domain (ECD) (aa 20 - 283) and a 95 aa cytoplasmic region.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought