Order Entry
ContactUsLinkComponent
Human Recombinant CD98 (from Cells)
Human Recombinant CD98 (from Cells)
# de catalogue 75789-656
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant CD98 (from Cells)
# de catalogue 75789-656
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    Lymphocyte Activation Antigen 4F2 Large Subunit|4F2 Heavy Chain Antigen|4F2 Cell-Surface Antigen Heavy Chain|MDU1|CD98|4F2hc|SLC3A2
  • Protein/peptide name:
    CD98
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    47.89 kD
  • Sequence:
    Arg206-Ala630
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 5% Threhalose, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-656
  • Supplier No.:
    91-490

Spécifications

A propos de cet article

CD98 is a single-pass type I I membrane protein which belongs to the SLC3A transporter family. SLC3A2/MDU1 is expressed ubiquitously in all tissues tested with highest levels detected in kidney, placenta and testis and weakest level in thymus. It consists of an 85 kDa glycosylated type II transmembrane heavy chain and a 40-50 kDa non-glycosylated light chain with 12 transmembrane segments. The heavy chain (SLC3A2) pairs with one of several light chains (SLC7A5, 6, 7, 8, 10, or 11) and is required for the cell surface expression and amino acid transport function of the light chains. It is involved in guiding and targeting of LAT1 and LAT2 to the plasma membrane. It also mediates integrin signaling, T cell costimulation, B cell proliferation, and viral fusion with cell membranes.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought