Order Entry
ContactUsLinkComponent
Human Recombinant TWSG1 (from Cells)
Human Recombinant TWSG1 (from Cells)
# de catalogue 75789-394
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant TWSG1 (from Cells)
# de catalogue 75789-394
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    TSG|TWSG1|Twisted Gastrulation Protein Homolog 1
  • Protein/peptide name:
    TWSG1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    23.18 kD
  • Sequence:
    Cys26-Phe223
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological Assays
  • Cat. no.:
    75789-394
  • Supplier No.:
    91-359

Spécifications

A propos de cet article

Twisted Gastrulation Protein Homolog 1 (TWSG1) is a 22 kDa secreted protein that belongs to the twisted gastrulation protein family. Human TWSG1 is synthesized as a 223 aa precursor that contains a 25 aa signal peptide and a 198 aa mature chain. TWSG1 may be involved in dorsoventral axis formation. TWSG1 seems to antagonize BMP signaling by forming ternary complexes with CHRD and BMPs, thereby preventing BMPs from binding to their receptors.TWSG1 can inhibit BMP activity by binding directly to BMP proteins, and can act the anti-BMP function, partly mediated by cleavage and degradation of CHRD, which releases BMPs from ternary complexes. TWSG1 may be an important modulator of BMP-regulated cartilage development, chondrocyte differentiation and thymocyte development.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought