Order Entry
ContactUsLinkComponent
Human Recombinant Biglycan (from Cells)
Human Recombinant Biglycan (from Cells)
# de catalogue 75789-382
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant Biglycan (from Cells)
# de catalogue 75789-382
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    SLRR1A|BGN|Bone/Cartilage Proteoglycan I|Biglycan|PG-S1
  • Protein/peptide name:
    Biglycan
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    40.47 kD
  • Sequence:
    Glu20-Lys368
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological Assays
  • Cat. no.:
    75789-382
  • Supplier No.:
    91-353

Spécifications

A propos de cet article

Biglycan is a 200-350 kD proteoglycan consisting of a 45 kD core protein and two chrondroitin/dermatan sulfate glycosaminoglycan chains. Biglycan binds to TGF-ß. It also binds to collagen type I in low ionic strength (less than 3 mM phosphate) buffer. At higher ionic strengths, Biglycan does not bind to collagen type I. It enhances the inhibition effect of TGF-ß on osteoclast proliferation at a concentration of 4-20 mg/ml. It also prevents the attachment of CHO cells to fibronectin, with a 50% inhibition at 17-21 mg/ml.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought