Order Entry
ContactUsLinkComponent
Human Recombinant Kallikrein 11 (from Cells)
Human Recombinant Kallikrein 11 (from Cells)
# de catalogue 75789-340
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant Kallikrein 11 (from Cells)
# de catalogue 75789-340
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    Trypsin-like Protease|KLK11|PRSS20|Hippostasin|Serine Protease 20|TLSP|hK11|Kallikrein-11
  • Protein/peptide name:
    Kallikrein 11
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    26.28 kD
  • Sequence:
    Glu19-Asn250
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, 2mM CaCl2, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-340
  • Supplier No.:
    91-332

Spécifications

A propos de cet article

Human Kallikrein 11 (KLK11) is a member of tissue kallikrein family which are extracellular serine proteases consisting of 15 members. Two isoforms of KLK11 are differentially expressed. Isoform 1 is predominantly expressed in brain and isoform 2 is preferentially expressed in prostate. Isoform 1 consists of a signal peptide,a short pro peptide and the mature chain.Isoform 2 contains an extra 32 amino acid N terminal to full-length isoform 1.KLK11 is a novel marker for ovarian and prostate cancer carcinomas. KLK11 can be activated by thermolysin and is active against a thioester substrate.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought