Order Entry
ContactUsLinkComponent
Human Recombinant Fetuin B (from Cells)
Human Recombinant Fetuin B (from Cells)
# de catalogue 75789-314
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant Fetuin B (from Cells)
# de catalogue 75789-314
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    16G2|FETUB|Fetuin-B|Gugu|Fetuin-like Protein IRL685
  • Protein/peptide name:
    Fetuin B
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    41.53 kD
  • Sequence:
    Cys16-Pro382
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-314
  • Supplier No.:
    91-319

Spécifications

A propos de cet article

Fetuin-B is a member of the Fetuin family that is part of the Cystatin superfamily of Cysteine Protease inhibitors. It is reported that Fetuin-B is highly expressed in liver tissue, in tongue and placenta tissues. Fetuin-B is a paralogue of Fetuin-A. Fetuin-A and Fetuin-B display similarities and differences in their characteristics, however, they share only 20% amino acid sequence identity. The amounts of Fetuin-B in human serum, unlike Fetuin-A, vary with gender and are higher in females than in males. Fetuin-B is an inhibitor of basic calcium phosphate precipitation but is less active than Fetuin-A. Fetuin-B expression is decreased in Fetuin-A deficient knock-out mice. The expression of Fetuin-B has been shown to be regulated by FXR (Farnesoid X Receptor), a nuclear receptor activated by bile acids. Evidence has shown that overexpression of Fetuin-B in skin squamous carcinoma cells suppresses tumor growth in nude mice. The function of Fetuin B is still not fully characterized.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought