Order Entry
ContactUsLinkComponent
Human Recombinant BMP Receptor II (from Cells)
Human Recombinant BMP Receptor II (from Cells)
# de catalogue 75789-242
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant BMP Receptor II (from Cells)
# de catalogue 75789-242
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    Bone Morphogenetic Protein Receptor Type-2|BMPR-II|PPH1|BMP Type-2 Receptor|Bone Morphogenetic Protein Receptor Type II|BMP Type II Receptor|BMPR-2|BMPR2
  • Protein/peptide name:
    BMP Receptor II
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    15.05 kD
  • Sequence:
    Ser27-Ile151
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-242
  • Supplier No.:
    91-283

Spécifications

A propos de cet article

Bone Morphogenetic Protein Receptor II (BMPR-II) is a Type II Serine/Threonine Kinase that mediates cellular responses to BMPs. BMPR-II is characterized by lacking of a GS domain, and presence of a C-terminal extension typical of type II receptors. BMPRII binds BMP2, BMP4 and BMP7 weakly in the absence of type I receptor, and the binding can be facilitated by the presence of the type I receptor, including BMPR-IA/Brk1, BMPR-IB, and ActR-I. BMPR-II plays a key role in cell growth. Defects in BMPR-II have been linked to primary pulmonary hypertension. Human and mouse BMPR-II are highly conserved and share 97 % amino acid sequence identity.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought