Order Entry
ContactUsLinkComponent
Human Recombinant Heat Shock beta 11 (from E. coli)
Human Recombinant Heat Shock beta 11 (from E. coli)
# de catalogue 75789-178
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant Heat Shock beta 11 (from E. coli)
# de catalogue 75789-178
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    Heat Shock Protein Beta-11|C1orf41|PP25|Placental Protein 25|Hspb11
  • Protein/peptide name:
    Heat Shock beta 11
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    18.5 kD
  • Sequence:
    Met1-Ser144
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 100mM NaCl, 2mM DTT, 10% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-178
  • Supplier No.:
    91-231

Spécifications

A propos de cet article

Heat Shock Protein beta -11 (HSPB11) is a stress-responsive protein that is required to deal with proteotoxic stresses. HSPB11 is composed of an IFT complex B composed of IFT88, IFT57, TRAF3IP1, IFT52, IFT27, HSPB11 and IFT20 and is detected in placenta. HSPB11 has beeb shown to form oligomeric complexes and prevent the aggregation of in vitro denaturated aldolase and glyceraldehyde-3-phosphate dehydrogenase in accordance with the chaperone model of HSPB1 and HSPB5. HSPB11 overexpression protected against etoposide-induced cell death that correlated with a decreased release of mitochondrial Cytochrome C into the cytosol. Inhibition of HSP90 function completely abrogated the protective effect of HSPB11. This data suggests that at least in the case of HSPB11, interaction with other chaperone machines besides HSPA1A may contribute to functional specificity and cellular functioning.

  • Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought