Order Entry
ContactUsLinkComponent
Human Recombinant BMP2 (from E. coli)
Human Recombinant BMP2 (from E. coli)
# de catalogue 75788-744
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant BMP2 (from E. coli)
# de catalogue 75788-744
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Biological activity:
    Recombinant BMP-2 is fully biologically active when compared to standards.<br />ED50 is less than 50 ng/ml as determined by the cytolysis of MC3T3-E1 cells.<br />Specific Activity of 2.0 x 10<sup>4</sup> IU/mg
  • Protein synonyms:
    BMP-2|BMP2A|Bone Morphogenetic Protein 2|BMP2|BMP-2A|Bone Morphogenetic Protein 2A
  • Protein/peptide name:
    BMP2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    13.3 kD
  • Sequence:
    Gln283-Arg396
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 50mM HAc . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in 50mM Acetic acid. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological assays
  • Cat. no.:
    75788-744
  • Supplier No.:
    91-010

Spécifications

A propos de cet article

Bone Morphogenetic Protein-2 (BMP-2) is one of the bone-growth regulatory factors that belong to the transforming growth factor-beta (TGF-beta) superfamily of proteins. BMPs are synthesized as large precursor molecules, which are cleaved by proteolytic enzymes. The active form of BMP-2 can consist of a dimer of two identical proteins or a heterodimer of two related bone morphogenetic proteins.

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought