Nalgene™ HDPE Tamper Evident Closures, Thermo Scientific
# de catalogue 89400-736
Fournisseur: Thermo Fisher Scientific
Spécifications
- Cap Size:38-430 mm
- Sterility:Sterile
- Description:Tamper-Evident Closure, Bulk-Pack
- Material:HDPE
- Cat. no.:89400-736
- Cap Color:Natural
- Supplier No.:342141-0384
Spécifications
A propos de cet article
Nalgene™ HDPE Tamper Evident Closures are specially designed for use with PET/PETG square media bottles.
- Available in 38-430 finish
- Natural HDPE closures are ADFC (Animal Derived Component-Free)
- Fits Nalgene PET and PETG Media Bottles with 38-430 neck finish (except 125mL PETG Media Bottles)
- Leakproof Guarantee when used with Nalgene PET and PETG Media Bottles
- Easy application and breaking of the tamper evident seal ring
- Gamma irradiated to provide 106 Sterility Assurance Level (SAL)
- Flat top surface for possible printing or labeling
Customers Who Bought This Also Bought