Two Faced Glove Box Holders, TrippNT
Fournisseur: TrippNT
Certificats
A propos de cet article
TrippNT's Two Faced Glove Box Holders install face-forawrd or face-back for two convenient choices to easily access gloves
Made of clear, high quality, tough PETG plastic, this holder is available in single, double, and triple verisions. Installs with keyholes and included hardware. Made in USA.
Customers Who Bought This Also Bought