Sliding Drawer Freezer Racks for Tubes, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Maximize tube storage capacity per freezer with Thermo Scientific™ sliding drawer racks for tubes (four and five inner door freezers), a racking solution for five inner-door, ultra-low temperature freezers.
Thermo Scientific™ freezer racking and tubing solutions offer a convenient, streamlined process to maximize sample storage in your laboratory. Racking solutions are designed to hold tubes ranging from 200 µL up to 5 mL, offering maximized sample storage capacity for both five and four inner door freezers.
Customers Who Bought This Also Bought