A propos de cet article
The Microalbumin Reagent Set is for the quantitative determination of low levels of albumin in urine by immunoturbidimetric assay.
- Quantitative determination of low levels of albumin in urine
- Calibrator for quantitating albumin in urine specimens
- Controls monitor methods to validate quantitation of samples.
For the quantitative determination of low levels of albumin in urine by immunoturbidimetric assay
This Microalbumin Multi-Calibrator Set is intended to be used for the calibration of Pointe Scientific’s Microalbumin immunoturbidimetric assay for quantitating albumin in urine specimens. For in vitro diagnostic use.
The Urine Microalbumin Controls are intended as a means of monitoring various microalbumin assay methods to validate quantitation of patient samples. Control materials having known component concentrations are an integral part of diagnostic procedures. Daily monitoring of control values establishes intralaboratory parameters for accuracy and precision of the test method.
For in vitro diagnostic use. When a sample is mixed with anti-human albumin goat antiserum, agglutination is caused by the antigen-antibody reaction. The turbidity is measured at 340nm and 700nm and albumin in the sample is quantitatively determined.
Calibrators: Microalbumin Multi-Calibrator Set, 6 Calibrators; Approx. values:
0, 0.5, 1.0, 5.0, 10.0, 30.0 mg/dl and control urines, both normal and abnormal.