Polytron® Homogenizer PT 2500 E, Kinematica
# de catalogue 97057-700
Fournisseur: Kinematica
Small Business Enterprise
Spécifications
- Electrical:115V
- Speed range:Up to 30,000 rpm
- Cat. no.:97057-700
- Volume Range:0.05 mL to 2000 mL
- Dimensions:Homogenizer
- Supplier No.:11010058
Spécifications
A propos de cet article
Kinematica Polytron® Homogenizer PT 2500 E Benchtop unit that uses the same dispersing generators as our handheld units for maximum versatility
3,5,7,12 and 20 mm generators for volumes from .05 to 2000 ml. Powerful 500 Watt motor with digital speed readout for reproducible results. 500-30,000 rpm.
Certifications: CE marked.
Customers Who Bought This Also Bought