StableTemp™ High-Temperature Heating Tapes, Cole-parmer®
Supplier: Argos Technologies
Certificates
About this item
Nongrounded, Heavy-duty insulated design.
- Maximum exposure temperature of 1400 ºF (760 ºC)
- Choose from power leads on same end or on opposite ends for easier wrapping
The 120 VAC tapes include a plug and the 240 VAC tapes are supplied with bare leads. The 120 VAC tapes with an opposite-end lead configuration are supplied with a half plug on each end; the half plugs fit together and connect to form a standard 120 VAC two-prong plug.
Customers Who Bought This Also Bought