Bio-Cane™ Cane and Canister Systems, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Bio-Cane™ cryostorage is ideal to store cells, tissues and biologicals in an economical and biological manner.
- Six stainless-steel canisters to accommodate vials
- Canisters are numbered and color-coded to simplify sample identification
- Durable aluminum construction and vacuum insulation
- Secure locking clasp and narrow-mouth design minimizes LN2 evaporation
Certifications: CE marked.
Spécifications
- Catalog No:
- 55709-984
- 55709-986
- 55709-982
- Model:
- Bio-Cane 34
- Bio-Cane 47
- Bio-Cane 20
- Evaporation rate:
- 0.18 L
- 0.39 L
- 0.09 L
- Height with lid:
- 67.6 cm (265/8")
- 67.3 cm (261/2")
- 65.3 cm (257/10")
- Neck int.Ø:
- 8.8 cm (31/2")
- 12.7 cm (5")
- 5.1 cm (2")
- Static holding time (days):
- 193 days
- 121 days
- 227 days
- Vial capacity:
- 630
- 1320
- 150