About this item
Spectroquant® Prove is the perfect tool for water analysis. It unites the simplicity you want with the security you need, and durability you expect.
- Simple and durable
- Intuitive controls
- Preprogrammed applications
- Long-life lamp
- Smart screen
- Custom measurements
The intuitive controls of the Spectroquant® Prove and the popular Spectroquant® test kit make analysis smoother than ever.
Some of the features of the Spectroquant® Prove spectrophotometers include Long-life Lamp, Smart Screen, Custom Measurements, Individualized Measuring Ranges, Cell Test Port, Robust Surfaces, Data Exchange, Reagent Tests, Easy Cleaning, Live ID, Preprogrammed Applications, Data on Demand, AQA Prime, and Matrix Check.
Spectroquant® Prove 100 is for routine applications and for those who primarily use our broad range of Spectroquant® test kits, or only perform Vis measurements. High quality and great value for money for your daily analyses.
Spectroquant® Prove 300 is for sensitive measurements. Thanks to its long-lasting xenon lamp, Prove 300 is ideal for more intensive use. What’s more, it is capable of both UV and Vis measurements, so you have greater flexibility for more intricate analyses.
Both the Prove 100 and 300 can accommodate 16 mm round cells, 10, 20 and 50 mm rectangular cells with automatic recognition. The Prove 600 can accommodate 16 mm round cells, 10, 20, 50 and 100 mm rectangular cells with automatic recognition.
Specifications
- Catalog No:
- EM1.73016.0001
- EM1.73017.0001
- EM1.73018.0001
- Interfaces:
- 2 x USB-A; 1 x USB-B
- 2 x USB-A; 1 x USB-B
- 2 x USB-A; 1 x USB-B
- Languages:
- 28
- 28
- 28