Briskheat® High-Temperature Heating Tapes, Thermo Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
These high-temperature heating tapes are made of resistance wires that are insulated with braided fiberglass knitted into a flat tape and covered with heavy fiberglass fabric
- Nongrounded
- Flexible
- Multipurpose
- Heavy-duty insulated design
- Maximum exposure temperature of 1400 ºF (760 ºC)
- Direct-contact heaters are ideal for use on conductive surfaces
Heating tapes are available with either a one-piece standard plug or a twopiece opposite-end power lead configuration. Opposite-end lead configuration provides increased flexibility when wrapping the tape around unique objects. The 120 VAC tapes include a plug and the 240 VAC tapes are supplied with bare leads. The 120 VAC tapes with an opposite-end lead configuration are supplied with a half plug on each end; the half plugs fit together and connect to form a standard 120 VAC
two-prong plug.
Customers Who Bought This Also Bought