Briskheat ® Silicone Rubber Heating Tapes with Adjustable Time Percentage Control, Cole-Parmer
Fournisseur: Argos Technologies
Certificats
A propos de cet article
These tapes allow users to regulate the length of time the heater is on and off.
- Heat to 260 °C (500 °F)
- Flexible silicone rubber is moisture resistant
- Tapes are chemical, abrasion, and moisture resistant
Solve a wide range of heating applications without risk of damage due to chemical and moisture spillover. Tapes are available with built-in time percentage control. Tapes are made of fine-strand resistance wires that are double insulated with braided fiberglass and knitted into flat tapes; then completely enclosed in high strength, high temperature-resistant silicone rubber. These tapes are ideal for tight radius heating applications and are chemical, abrasion, and moisture resistant.
Caution: Heating tapes should not be overlapped.
Customers Who Bought This Also Bought