AlphaTec™ 56-015 PVC Aprons, Ansell
Fournisseur: Ansell Healthcare
Certificats
A propos de cet article
Wherever oil, grease and chemicals are present, Ansell aprons help workers keep clean and protected from harmful contact.
- Excellent splash protection
- Superior flexibility
- Adjustable ties ensure optimal fit
These medium-duty aprons feature 5- to 6-millimeter die-cut PVC and offer users a combination of chemical splash protection and flexible comfort
Use in the food processing, food service, and light manufacturing industries. Uses include food handling and preparation, industrial maintenance, and chemical processing.
The one-piece construction is economical, and features integral head opening and ties. Seamless vinyl construction minimizes opportunity to trap contaminants and offers resistance to puncture and abrasions.
Spécifications
- Catalog No:
- 80093-264
- Garment application:
- Food, Industrial, Chemical protection
- Thickness:
- 5-6 mils