AlphaTec™ 56-007 PVC Aprons, Ansell
Fournisseur: Ansell Healthcare
Certificats
A propos de cet article
Wherever oil, grease and chemicals are present, Ansell aprons help workers keep clean and protected from harmful contact.
- Excellent splash protection
- Superior flexibility
- Adjustable ties ensure optimal fit
These medium-duty aprons feature 5- to 6-millimeter PVC and have a sewn edge finish
Use in the food processing, food service, and chemical processing industries. Uses include food handling and preparation, industrial maintenance, and chemical processing.
They provide superior flexibility and excellent resistance to punctures, abrasions, oil, fat, and chemicals. PVC grommets are permanently sealed and 3-piece 28" ties provide a durable, consistent and adjustable fit. Bib styles in four sizes with adjustable ties ensure optimal fit for a wide range of users.
Spécifications
- Catalog No:
- 80093-226
- 80093-222
- Garment application:
- Food, Industrial, Chemical protection, Oils, fats and fuels
- Food, Industrial, Chemical protection, Oils, fats and fuels
- Thickness:
- 5-6 mils
- 5-6 mils