Metal Hub Needles, Hamilton
Fournisseur: Hamilton
Certificats
A propos de cet article
Metal hub (nickel plated brass) needles can be used with TLL syringes and LT or TLL connectors.
- Autoclavable
- 304 stainless steel needles
- Custom needle lengths and point styles are available
Packaging: Conveniently packed with 6 needles per pack.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.