SpecMat-H Anti-Static Mats, Precut, 2.54 mm Thickness, ACL
Fournisseur: ACL Staticide
Certificats
A propos de cet article
SpecMat-H Mats are static dissipative, single-layer, ESD-safe thermoplastic vinyl mats made for bench tops and other work surfaces
These single layer mats include two 10mm female snaps at top and bottom left side. Snaps can be located elsewhere if specified.
Available in 8 standard sizes. Custom sizes are also available. Mats are 100% recyclable, and made in the USA.
They also work as an excellent shelving liner. Lifetime warranty on electrical resistance (10⁸ to 10⁹ ohms).
Certifications: Compliant to ANSI/ESDA S20.20.
Customers Who Bought This Also Bought