Kel-F® Hub Needles, Hamilton
Supplier: Hamilton
Certificates
About this item
Kel-F® hub (KF) needles can be used with Luer Tip (LT) and PTFE Luer Lock (TLL) syringes and connectors.
- Autoclavable, gas sterilizable
- 304 stainless steel needles
Packaging: Conveniently packed with 6 needles per pack.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.