About this item
Homogenizers are used to rapidly break down plant and animal tissue for chemical analysis, prepare emulsions and samples for titrations, and extract pesticides in soil samples
The PT 10-35 GT with its 1200 watt motor can be used for applications with operating volumes up to 10 liters. More than 30 dispersing generator with different geometries, designs, sizes, materials and coatings are available. The electronic control system continuously compares the preselected rpm speed with the actual speed and adjusts it so that the speed remains constant even as the viscosity increases.
EasyCare™ generators disassemble for cleaning, maintenance, and sterilization, making them ideal for use when cross-contamination is a concern and cleaning is necessary. The generators are also designated "Quick-Connect" as they can be connected/disconnected from the homogenizer quickly and easily. These generators work via the rotor/stator system, in which samples are drawn into the center of the generator tip, vigorously mixed, accelerated, pressed through the narrow gap between the rotor and stator, and forced out between the teeth of the stator.
Provide fast, reproducible homogenization, emulsion, dispersion, and mixing, with consistent sample-to-sample uniformity. These units can also be used to homogenize heart, liver, uterus, and smooth muscle for the isolation of mitochondria, intact golgi apparatus, and nuclei, as well as grains and seeds for nutritional analysis.
Specifications
- Catalog No:
- 97036-086
- Speed range:
- 30000 rpm max.
- Dimensions:
- Ø: 5-36 mm Generator
- Power rating:
- 1200W