ContactUsLinkComponent
Stainless Steel Student Weight Set
Stainless Steel Student Weight Set
# de catalogue 10754-334
Fournisseur:  Troemner
Product available on GSA Advantage®
Economical Stainless Steel 100 g x1 g Weight Set Class 7, For rough weighing operations
Stainless Steel Student Weight Set
# de catalogue 10754-334
Fournisseur:  Troemner
Product available on GSA Advantage®
Spécifications
  • Description:
    Stainless Steel Cylinder Weight Sets, 1 to 100g
  • Cat. no.:
    10754-334
  • Supplier No.:
    311S
Spécifications
A propos de cet article
Set of 9 cylinder weights.
  • Student set
The Set of 9 cylinder weights includes a 100 g, 50 g, two 20 g, 10 g, 5 g, two 2 g, 1 g weight.
Customers Who Bought This Also Bought