VWR® Graduated Wide Mouth AC Round Bottles, Clear
Fournisseur: Avantor
Certificats
A propos de cet article
VWR® Clear, graduated, wide mouth AC round bottles are manufactured from soda-lime glass that conforms to USP Type III requirements.
- Soda-lime glass fabrication
- Graduated
- Wide mouth
Bottles are available in three packaging options. They may be purchased in convenience packs with closures attached to help maintain cleanliness, in bulk packs with unattached closures in separate bags, or bulk packed without closures.
Customers Who Bought This Also Bought