VWR® Phenolic Resin Worksurfaces for Carts, Black
Fournisseur: VWR International
Certificats
A propos de cet article
Phenolic resin worksurfaces are composed of cellulose fiber-reinforced, phenolic resin core with a highly crosslinked, polyurethane, copolymer surface.
- Each worksurface has three square edges and one edge with a ¹/₈" chamfer
- For High-Column worksurfaces, two or three cut-outs accomodate 76" high columns, depending on bench width
- High-Column worksurfaces may be used with sitting/bench height carts when height-adjustment is required
Phenolic resin worksurfaces are chemical-resistant and non-flammable.
Customers Who Bought This Also Bought